Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502106.2 | XP_003502154.1 | 2 - 3 | 382 | 77 | |
GenBank mRNA | JI890581.1 | - | 2 - 3 | 385 | 77 | |
JP059383.1 | - | 2 - 3 | 385 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |