Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646136.1 | XP_007644326.1 | 3 - 4 | 1657 | 96 | |
GenBank mRNA | JI896377.1 | - | 1 - 2 | 511 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Inventoried Probe spans exons |