Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655186.1 | XP_007653376.1 | 2 - 3 | 310 | 97 | |
GenBank mRNA | JI865827.1 | - | 2 - 3 | 250 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |