Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641394.1 | XP_007639584.1 | 1 - 2 | 960 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |