Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646422.1 | XP_007644612.1 | 2 | 2894 | 99 | |
XM_007646423.1 | XP_007644613.1 | 2 | 2918 | 99 | ||
XM_007646424.1 | XP_007644614.1 | 2 | 2916 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |