Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512067.1 | XP_003512115.1 | 3 | 1244 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |