Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505755.1 | XP_003505803.1 | 1 | 921 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Both primers and probe map within a single exon |