Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654695.1 | XP_007652885.1 | 1 | 1221 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Both primers and probe map within a single exon |