Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043629.1 | XP_017899118.1 | 3 - 4 | 695 | 101 | |
GenBank mRNA | JT737444.1 | - | 3 - 4 | 314 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Capra hircus Probe spans exons |