Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063156.1 | XP_017918645.1 | 1 - 2 | 1127 | 92 | |
XM_018063157.1 | XP_017918646.1 | 1 - 2 | 1131 | 92 | ||
GenBank mRNA | JO440828.2 | - | 1 - 2 | 439 | 92 | |
KA346148.1 | - | 1 - 2 | 993 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |