Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062728.1 | XP_017918217.1 | 3 - 4 | 1443 | 67 | |
XM_018062729.1 | XP_017918218.1 | 3 - 4 | 1188 | 67 | ||
XM_018062730.1 | XP_017918219.1 | 3 - 4 | 1070 | 67 | ||
GenBank mRNA | JO503707.2 | - | 1 - 2 | 560 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |