Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H19orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051015.1 | XP_017906504.1 | 3 - 4 | 501 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |