Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698536.3 | XP_005698593.1 | 4 - 5 | 488 | 78 | |
XM_018041710.1 | XP_017897199.1 | 5 - 6 | 467 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |