Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H8orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051849.1 | XP_017907338.1 | 2 - 3 | 267 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |