Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050331.1 | XP_017905820.1 | 3 - 4 | 560 | 82 | |
GenBank mRNA | JT639803.1 | - | 3 - 4 | 363 | 82 | |
JT759829.1 | - | 2 - 3 | 299 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |