Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683150.3 | XP_005683207.1 | 2 - 3 | 997 | 96 | |
GenBank mRNA | JO395104.2 | - | 2 - 3 | 510 | 96 | |
KA346166.1 | - | 2 - 3 | 281 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |