Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680026.3 | XP_005680083.1 | 2 - 3 | 269 | 67 | |
XM_005680027.3 | XP_005680084.1 | 2 - 3 | 273 | 67 | ||
GenBank mRNA | JO643034.2 | - | 2 - 3 | 958 | 67 | |
JT732935.1 | - | 3 - 4 | 234 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |