Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013976300.2 | XP_013831754.1 | 4 - 5 | 1353 | 67 | |
XM_018044288.1 | XP_017899777.1 | 4 - 5 | 637 | 67 | ||
GenBank mRNA | JO622537.2 | - | 1 - 2 | 1390 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |