Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680816.3 | XP_005680873.2 | 1 - 2 | 1030 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Endogenous Controls Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |