Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043532.1 | XP_017899021.1 | 11 - 12 | 1425 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |