Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678639.3 | XP_005678696.2 | 2 - 3 | 595 | 71 | |
XM_018042342.1 | XP_017897831.1 | 5 - 6 | 971 | 71 | ||
XM_018042352.1 | XP_017897841.1 | 5 - 6 | 1365 | 71 | ||
GenBank mRNA | JT664877.1 | - | 2 - 3 | 1436 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |