Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051885.1 | XP_017907374.1 | 6 - 7 | 856 | 79 | |
GenBank mRNA | KA329469.1 | - | 6 - 7 | 818 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |