Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966452.2 | XP_013821906.2 | 3 - 4 | 736 | 74 | |
XM_018053101.1 | XP_017908590.1 | 6 - 7 | 1170 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |