Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066939.1 | XP_017922428.1 | 4 - 5 | 693 | 58 | |
GenBank mRNA | JO359473.2 | - | 1634 | 58 | ||
JO579201.1 | - | 5 - 6 | 3435 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |