Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972132.2 | XP_013827586.2 | 1 - 2 | 2043 | 89 | |
XM_018064780.1 | XP_017920269.1 | 1 - 2 | 2043 | 89 | ||
GenBank mRNA | JO427335.1 | - | 1 - 2 | 705 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |