Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UROD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678547.3 | XP_005678604.2 | 6 - 7 | 766 | 82 | |
XM_005678549.3 | XP_005678606.2 | 5 - 6 | 676 | 82 | ||
GenBank mRNA | JO550986.2 | - | 3 - 4 | 452 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |