Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043756.1 | XP_017899245.1 | 5 - 6 | 1505 | 65 | |
XM_018043757.1 | XP_017899246.1 | 4 - 5 | 1367 | 65 | ||
GenBank mRNA | JT662518.1 | - | 4 - 5 | 4242 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |