Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698746.2 | XP_005698803.1 | 1 - 2 | 407 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |