Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690895.3 | XP_005690952.1 | 2 - 3 | 1166 | 88 | |
XM_013970438.2 | XP_013825892.1 | 2 - 3 | 1150 | 88 | ||
XM_018060613.1 | XP_017916102.1 | 2 - 3 | 1010 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |