Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102177222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044173.1 | XP_017899662.1 | 2 - 3 | 182 | 76 | |
GenBank mRNA | JO520060.2 | - | 4 - 5 | 494 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |