Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058329.1 | XP_017913818.1 | 2 - 3 | 682 | 105 | |
GenBank mRNA | KU643198.1 | - | 2 - 3 | 282 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |