Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059366.1 | XP_017914855.1 | 7 - 8 | 1041 | 75 | |
XM_018059367.1 | XP_017914856.1 | 7 - 8 | 1046 | 75 | ||
GenBank mRNA | JT743526.1 | - | 3 - 4 | 879 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |