Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287178.1 | NP_001274107.1 | 4 - 5 | 371 | 99 | |
XM_005678573.3 | XP_005678630.2 | 5 - 6 | 528 | 99 | ||
XM_018039364.1 | XP_017894853.1 | 6 - 7 | 595 | 99 | ||
XM_018039370.1 | XP_017894859.1 | 5 - 6 | 487 | 99 | ||
XM_018039373.1 | XP_017894862.1 | 6 - 7 | 565 | 99 | ||
GenBank mRNA | JO610506.2 | - | 3 - 4 | 1032 | 99 | |
KF549985.1 | - | 4 - 5 | 371 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |