Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287236.1 | NP_001274165.1 | 2 - 3 | 474 | 65 | |
GenBank mRNA | KC119191.1 | - | 2 - 3 | 474 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |