Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285736.1 | NP_001272665.1 | 2 - 3 | 338 | 87 | |
XM_018063565.1 | XP_017919054.1 | 3 - 4 | 1414 | 87 | ||
XM_018063566.1 | XP_017919055.1 | 3 - 4 | 1414 | 87 | ||
GenBank mRNA | GU441534.1 | - | 2 - 3 | 338 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |