Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314186.1 | NP_001301115.1 | 11 - 12 | 1952 | 65 | |
XM_018063571.1 | XP_017919060.1 | 11 - 12 | 1969 | 65 | ||
GenBank mRNA | KF803258.1 | - | 11 - 12 | 1952 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |