Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285587.1 | NP_001272516.1 | 3 - 4 | 147 | 102 | |
GenBank mRNA | JT653927.1 | - | 3 - 4 | 102 | ||
X60763.1 | - | 3 - 4 | 147 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |