Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285588.1 | NP_001272517.1 | 1 - 2 | 335 | 67 | |
GenBank mRNA | EU847284.1 | - | 1 - 2 | 335 | 67 | |
EU847289.1 | - | 1 - 2 | 335 | 67 | ||
EU888137.1 | - | 1 - 2 | 335 | 67 | ||
JF824148.1 | - | 1 - 2 | 335 | 67 | ||
JF824149.1 | - | 1 - 2 | 335 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |