Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285595.1 | NP_001272524.1 | 7 - 8 | 1057 | 89 | |
GenBank mRNA | HQ846826.1 | - | 7 - 8 | 1057 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |