Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285608.1 | NP_001272537.1 | 1 - 2 | 290 | 71 | |
XM_013963900.1 | XP_013819354.1 | 1 - 2 | 355 | 71 | ||
GenBank mRNA | EF035041.1 | - | 1 - 2 | 290 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |