Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285698.1 | NP_001272627.1 | 2 - 3 | 206 | 72 | |
GenBank mRNA | FJ594753.1 | - | 2 - 3 | 206 | 72 | |
JO580480.1 | - | 5 - 6 | 805 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |