Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285759.1 | NP_001272688.1 | 1 - 2 | 114 | 69 | |
GenBank mRNA | AY389343.1 | - | 1 - 2 | 114 | 69 | |
EF199763.1 | - | 1 - 2 | 77 | 69 | ||
GQ369727.1 | - | 1 - 2 | 65 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |