Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286442.1 | NP_001273371.1 | 1 - 2 | 222 | 84 | |
GenBank mRNA | AF276985.1 | - | 1 - 2 | 54 | 84 | |
AY304502.1 | - | 1 - 2 | 183 | 84 | ||
D86587.1 | - | 1 - 2 | 222 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |