Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700421.2 | XP_005700478.2 | 2 - 3 | 927 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |