Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691577.3 | XP_005691634.2 | 4 - 5 | 382 | 107 | |
XM_005691578.3 | XP_005691635.2 | 4 - 5 | 378 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |