Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709376.3 | XP_005709433.1 | 1 - 2 | 186 | 104 | |
GenBank mRNA | JT632711.1 | - | 1 - 2 | 175 | 104 | |
JT650680.1 | - | 1 - 2 | 173 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |