Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048110.1 | XP_017903599.1 | 4 - 5 | 747 | 67 | |
GenBank mRNA | JO449234.2 | - | 4 - 5 | 610 | 67 | |
JT705662.1 | - | 4 - 5 | 469 | 67 | ||
JT717704.1 | - | 5 - 6 | 539 | 67 | ||
KA313250.1 | - | 4 - 5 | 548 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |