Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102172074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056816.1 | XP_017912305.1 | 2 - 3 | 3038 | 71 | |
GenBank mRNA | JO648731.2 | - | 2 - 3 | 231 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |