Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690417.2 | XP_005690474.1 | 5 - 6 | 565 | 103 | |
XM_018060098.1 | XP_017915587.1 | 6 - 7 | 628 | 103 | ||
XM_018060099.1 | XP_017915588.1 | 6 - 7 | 629 | 103 | ||
XM_018060101.1 | XP_017915590.1 | 5 - 6 | 562 | 103 | ||
XM_018060102.1 | XP_017915591.1 | 5 - 6 | 547 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |