Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967643.2 | XP_013823097.2 | 4 - 5 | 457 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |